MilliporeSigma™ Opticap™ Sterile XL150 Capsule Filters
For sterilizing grade performance and mycoplasma removal
Supplier: MilliporeSigma™ KHGES015FF3
Description
- High-flux hydrophilic sterilizing-grade 0.1μm rated PES membrane for improved process economy
- On-board PES membrane prefilter for extended throughput in high fouling media
- Robust construction
- Withstand multiple sterilization cycles
- Easy wetting and integrity testing
- Broad chemical compatibility across a wide pH range (pH 1–14)
- Hydrophilic Polyethersulfone filter
- Polyester supports
- Structural components: Gamma stable polypropylene/Polysulfone
- Silicone vent O-rings
- Gamma compatible sterilization
Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals
Specifications
0.5/0.2 μm | |
Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals | |
0.56 in. (14mm) hose barb | |
9.7 cm |
0.014 m2 | |
0.75 in. (19mm) sanitary flange | |
3.8 in. |