Please login to your online account to display your discounted pricing

EMD Millipore Nylon Woven Net Filters

Compatible with a broad range of solvents

$143.00 - $587.00

Specifications

Color Code White
Color (Overall) White
Product Type Net Filter
Feedwater Temperature (Metric) 100°C
Filter Type Plain
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Pore Density Pore Size Thickness (English) Price Quantity    

NY1100010

 
emd millipore
NY1100010
1.18 in. 6% 11μm 0.002 in. Pack for $565.00

NY1H04700

 
emd millipore
NY1H04700
1.8 in. 44% 100μm 0.003 in. Pack of 100 Each for $178.00

NY1104700

 
emd millipore
NY1104700
1.8 in. 6% 11μm 0.002 in. Pack of 100 Each for $178.00

NY2002500

 
emd millipore
NY2002500
0.98 in. 14% 20μm 0.002 in. Pack of 100 Each for $143.00

NY2004700

 
emd millipore
NY2004700
1.8 in. 14% 20μm 0.002 in. Pack of 100 Each for $178.00

NY2H04700

 
emd millipore
NY2H04700
1.8 in. 49% 120μm 0.003 in. Pack of 100 Each for $178.00

NY4102500

 
emd millipore
NY4102500
0.98 in. 31% 41μm 0.001 in. Pack of 100 Each for $143.00

NY3002500

 
emd millipore
NY3002500
0.98 in. 17% 30μm 0.002 in. Pack of 100 Each for $143.00

NY3004700

 
emd millipore
NY3004700
1.8 in. 17% 30μm 0.002 in. Pack of 100 Each for $178.00

NY6004700

 
emd millipore
NY6004700
1.8 in. 41% 60μm 0.001 in. Pack of 100 Each for $178.00

NY4H02500

 
emd millipore
NY4H02500
0.98 in. 43% 140μm 0.004 in. Pack of 100 Each for $143.00

NY6H04700

 
emd millipore
NY6H04700
1.8 in. 52% 160μm 0.003 in. Pack of 100 Each for $178.00

NY8002500

 
emd millipore
NY8002500
0.98 in. 40% 80μm 0.002 in. Pack of 100 Each for $143.00

NY8004700

 
emd millipore
NY8004700
1.8 in. 40% 80μm 0.002 in. Pack of 100 Each for $178.00

NY8H04700

 
emd millipore
NY8H04700
1.8 in. 47% 180μm 0.005 in. Pack of 100 Each for $178.00

NY1H00010

 
emd millipore
NY1H00010
1.18 in. 44% 100μm 0.003 in. Pack for $587.00

NY1H02500

 
emd millipore
NY1H02500
0.98 in. 44% 100μm 0.003 in. Pack of 100 Each for $143.00

NY4104700

 
emd millipore
NY4104700
1.8 in. 31% 41μm 0.001 in. Pack of 100 Each for $178.00
Description & Specifications

Specifications

Color Code White
Color (Overall) White
Product Type Net Filter
Feedwater Temperature (Metric) 100°C
Filter Type Plain
For Use With (Application) Particle filtration, particle analysis, prefiltration of solvents and paint monitoring
Hydrophobicity Hydrophilic
Material Nylon
Temperature (Metric) Operating 100°C

  • Hydrophilic
  • Plain surface
  • Sterilization: Gamma or EO compatible
  • Operating Temperature: 75°C max.
  • Color: white
  • Mesh openings range from 10 to 180μm
  • Ideal for collection of algae and cells, particle analysis, large particulate filtration, background filter for automated particle imaging systems, prefiltration of solvents, paint monitoring