Please login to your online account to display your discounted pricing

EMD Millipore Nylon Net Filters

$143.00 - $143.00

Specifications

Color (Overall) White
Diameter (English) 0.98 in.
Diameter (Metric) 2.5cm
Product Type Net Filter
Filter Type Plain
View More Specs
Products
Catalog Number Mfr. No. Pore Density Pore Size Price Quantity    

NY6002500

 
emd millipore
NY6002500
41% 60μm Pack of 100 Each for $143.00

NY8H02500

 
emd millipore
NY8H02500
47% 180μm Pack of 100 Each for $143.00
Description & Specifications

Specifications

Color (Overall) White
Diameter (English) 0.98 in.
Diameter (Metric) 2.5cm
Product Type Net Filter
Filter Type Plain
Hydrophobicity Hydrophilic
Material Nylon
Temperature (Metric) Operating 100°C