Please login to your online account to display your discounted pricing
EMD Millipore Nuts for Hibar™/LiChroCART™ Columns
Manufacturer: emd millipore  1512370001
 View more versions of this product
Catalog No. M1512370001
  • / Each
Description & Specifications
Specifications
Includes Two knurled nuts, six PVDF double cones
Quantity Not Applicable
For Use With (Equipment) Hibar columns/LiChroCART cartridges
Type Nuts
For Use With (Application) For capillary connection