EMD Millipore™ Nutrient Media
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
Manufacturer:  EMD Millipore™ MHA000P2E
 View more versions of this product
Catalog No. MHA000P2E
Description
Description
  • Each dry powder lot for media preparation assayed for purity and compared to satisfactory lot
  • Each m-Endo broth for total coliform lot and m-FC broth for fecal coliform analysis lot certified in accordance with the Safe Drinking Water Act
  • Lot QC certificates included in each package as added assurance for critical procedures
Order Info
Nutrient media comes in liquid form, packaged in 2mL plastic ampules. Refrigerate as detailed on label.
Specifications
Specifications
Pink
Liquid
m-Endo Total Coliform Broth
50 x 2mL
For testing drinking Water for total Coliform detection
Total Coliforms
United States
Ampule
2-10°C
Documents