Please login to your online account to display your discounted pricing

EMD Millipore Nutrient Media


Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs

Manufacturer: emd millipore  MHA000P2D

 View more versions of this product

Catalog No. MHA000P2D

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Packaging Ampule
Storage Requirements 2-10°C
For Use With (Application) Used for breweries to detect and enumerate bacteria present in small numbers in a mixed flora sample, Cycloheximide inhibits most Yeasts and molds
Format Liquid
Manufacturing Origin United States
Media Type Wallerstein Differential Broth
Quantity 50 x 2mL
Color Green
Detects Aciduric bacteria (Yeast and mold are inhibited)
Shelf Life 1 Year