missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Novagen™ Strep·Tag™ II Monoclonal Antibody
Mouse monoclonal antibody (IgG1) that has high specificity and affinity for 8-aa Strep·Tag II peptide
Supplier: MilliporeSigma™ 715903
Description
- Specificity: 8-aa Strep.Tag II peptide, N terminal
- Sensitivity: 5ng
- Species/Isotype: Mouse monoclonal IgG1
- Form: Lyophilized, should be reconstituted in PBS at a concentration of 0.2mg/mL
- Cross-reactivity: Negligible with bacterial, insect or mamallian cell lysates
- Application: Immunoblotting
- Working Dilution: 1:1000 for Immunoblotting
Specifications
Strep-Tag II Monoclonal Antibody | |
100 μg | |
-20°C |