Please login to your online account to display your discounted pricing
EMD Millipore Multiwell Plate with Collagen I Coating
Enables most adherent cells to attach and proliferate
$89.50 - $89.50
Specifications
Sterile No
Surface Coating Collagen I
For Use With (Application) Cell attachment, cell culture, cell differentiation, immunocytochemistry
Length (Metric) Exterior 12.8cm
Material Polystyrene
View More Specs
Products
Catalog Number Mfr. No. No. of Wells Price Quantity    
PICL24P05
emd millipore
PICL24P05
24 Pack of 5 Each for $89.50
Description & Specifications
Specifications
Sterile No
Surface Coating Collagen I
For Use With (Application) Cell attachment, cell culture, cell differentiation, immunocytochemistry
Length (Metric) Exterior 12.8cm
Material Polystyrene
Width (Metric) Exterior 8.55cm
6- and 24-well format polystyrene plates provide flexibility to run multiple samples simultaneously
  • Uses ECM proteins combined with cellular adhesion molecules on the surface of cells
  • Facilitates necessary substrate for many fastidious cells to attach to the surface
  • Allows efficient plating and higher proliferation rate for cells
  • Produces demonstrable enhanced differentiation potential in cells
  • Non-sterile
Individually blister packaged in packs of five