Please login to your online account to display your discounted pricing

EMD Millipore Multiwell Plate with Collagen I Coating


Enables most adherent cells to attach and proliferate

Manufacturer: emd millipore  PICL24P05

 View more versions of this product

Catalog No. PICL24P05

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Culture Area 1.9cm2
For Use With (Application) Cell attachment, cell culture, cell differentiation, immunocytochemistry
Format 24-well
Length (Metric) Exterior 12.8cm
Material Polystyrene
No. of Wells 24
Sterile No
Surface Coating Collagen I
Type 24-Well Multiwell Plate
Width (Metric) Exterior 8.55cm
Format 24-well
Wells 24
Material Polystyrene
Type 24-Well Multiwell Plate