Please login to your online account to display your discounted pricing

EMD Millipore MultiScreenHTS Solubility Filter Plate


96-well filter plate designed and optimized for aqueous solubility assays

Manufacturer: emd millipore  MSSLBPC10

 View more versions of this product

Catalog No. MSSLBPC10

  • / Pack of 10 Each
 

Description & Specifications

Specifications

No. of Wells 96
Sterile No
For Use With (Application) Aqueous solubility assays
Material Styrene
Type MultiScreenHTS-PCF Filter Plate
Membrane Polycarbonate
Color Clear
Pore Size 0.4μm

  • High-throughput system to classify or quantify the aqueous solubility of compound libraries stored in solvent as a concentrated solution
  • Analysis time in under four hours using <100μg of sample per test
  • Automation compatible, with space for bar coding
  • Validated for high drug recovery
  • Low-binding membrane and low-binding plate materials