missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore MultiScreenHTS Solubility Filter Plate

96-well filter plate designed and optimized for aqueous solubility assays

$526.00 - $2,267.00

Specifications

Color Clear
Material Styrene
Sterile No
Type MultiScreenHTS-PCF Filter Plate
For Use With (Application) Aqueous solubility assays
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MSSLBPC50 EMD Millipore
MSSLBPC50
Pack of 50 Each for $2,267.00
MSSLBPC10 EMD Millipore
MSSLBPC10
Pack of 10 Each for $526.00
Specifications

Description

  • High-throughput system to classify or quantify the aqueous solubility of compound libraries stored in solvent as a concentrated solution
  • Analysis time in under four hours using <100μg of sample per test
  • Automation compatible, with space for bar coding
  • Validated for high drug recovery
  • Low-binding membrane and low-binding plate materials
Description & Specifications

Specifications

Clear
No
Aqueous solubility assays
96
Styrene
MultiScreenHTS-PCF Filter Plate
Polycarbonate
0.4μm