Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™HTS 96-Well Filter Plates with Negatively Charged Membrane


Binds positive charge—ideal for kinase assays with peptide substrates and enzyme assays

Manufacturer: emd millipore  MSPHN6B50

 View more versions of this product

Catalog No. MSPHN6B50

  • / Pack of 50 Each
 

Description & Specifications

Specifications

For Use With (Application) Protein kinase assays with peptide substrates, enzyme assays, binds positive charge
Width (Metric) 8.55cm
Height (Metric) 1.44cm
Length (English) 0.5 in.
Sterile No
Width (English) 3.36 in.
Filtration Area 0.26cm2
Material Barex™/TiO2
Dimensions (L x W x H) 0.5 x 3.36 x 0.56 in. (1.28 x 8.55 x 1.44cm)
Color Opaque
Volume (Metric) Well 0.05 to 0.25mL
Height (English) 0.56 in.
Length (Metric) 1.28cm
No. of Wells 96
Certifications/Compliance ANSI, SBS
Pore Size 0.65μm
Membrane Negatively Charged Phosphocellulose
Type MultiScreenHTS-PH Plate