Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™HTS 96-Well Filter Plates with Negatively Charged Membrane


Binds positive charge—ideal for kinase assays with peptide substrates and enzyme assays

Manufacturer: emd millipore  MSPHN6B10

 View more versions of this product

Catalog No. MSPHN6B10

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Filtration Area 0.26cm2
Volume (Metric) Well 0.05 to 0.25mL
Dimensions (L x W x H) 0.5 x 3.36 x 0.56 in. (1.28 x 8.55 x 1.44cm)
Certifications/Compliance ANSI, SBS
Type MultiScreenHTS-PH Plate
Height (English) 0.56 in.
Material Barex™/TiO2
Pore Size 0.65μm
Length (English) 0.5 in.
Length (Metric) 1.28cm
Membrane Negatively Charged Phosphocellulose
Sterile No
Width (English) 3.36 in.
No. of Wells 96
Width (Metric) 8.55cm
Height (Metric) 1.44cm
Color Opaque
For Use With (Application) Protein kinase assays with peptide substrates, enzyme assays, binds positive charge