Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™HTS 96-Well Filter Plates with Negatively Charged Membrane

Binds positive charge—ideal for kinase assays with peptide substrates and enzyme assays

$267.00 - $1,201.00

Specifications

Color Opaque
Dimensions (L x W x H) 0.5 x 3.36 x 0.56 in. (1.28 x 8.55 x 1.44cm)
Material Barex™/TiO2
Sterile No
Width (Metric) 8.55cm
View More Specs
Products
Catalog Number Mfr. No. Quantity Type Price Quantity    

MSPHN6B10

 
emd millipore
MSPHN6B10
Pack of 10 Each for $267.00

MSPHN6B50

 
emd millipore
MSPHN6B50
Pack of 50 Each for $1,201.00
Description & Specifications

Specifications

Color Opaque
Dimensions (L x W x H) 0.5 x 3.36 x 0.56 in. (1.28 x 8.55 x 1.44cm)
Material Barex™/TiO2
Sterile No
Width (Metric) 8.55cm
Filtration Area 0.26cm2
For Use With (Application) Protein kinase assays with peptide substrates, enzyme assays, binds positive charge
Height (Metric) 1.44cm
Length (Metric) 1.28cm
Membrane Negatively Charged Phosphocellulose
No. of Wells 96
Volume (Metric) Well 0.05 to 0.25mL

  • Two-piece plate design eliminates surface contact with individual well “drip directors,” prevents crosstalk between wells, allows membrane access, and provides easy, contamination-free stacking and improved seal with vacuum manifolds
  • Extended skirt enables stacking directly with collection plates, protects recessed underdrain, provides ample surface for barcode labels, and improves vacuum seal
  • Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
  • Meet ANSI™/SBS 2004 plate specifications
  • Standardized dimensions allow full compatibility with automated gripper arms, stackers, barcode labels, plate washers, and plate readers; aligned, rigid sidewalls permit easy handling by robotic gripper arms
  • Opaque plastic to prevents light transfer between wells during direct scintillation counting