missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore MultiScreenHTS Durapore™ 96-Well Filter Plates

Designed for full automation compatibility. EMD Millipore MultiScreenHTS Durapore™ 96-Well Filter Plates are innovative plates ideal for bead based assays. Opaque Barex™ and clear, sterile or non-sterile styrene and racks are available in a range of pore sizes.

Manufacturer:  EMD Millipore MSGVS2210

 View more versions of this product

Catalog No. MSGVS2210


 

Specifications

Description

Filter Plates; Millipore; MultiScreenHTS 96 well, 0.22 μm Pore Size; Glass fiber/Hydrophilic PVDF; Styrene; Nonsterile; Durapore; For For Bead/rasin based; 50/Pack; Opaque

For use in receptor-ligand binding, general sample preparation, ELISPOT, enzyme activity, bead-based assays, whole organism visualization, and radioisotope detection.
  • Standardized dimensions and aligned, rigid sidewalls
    • Allows full compatibility with automated/robotic gripper arms, stackers, plate washers, bar code labels, and plate readers
  • Unique two-piece plate design eliminates surface contact with individual well “drip directors” to prevent crosstalk between wells
  • Extended skirt and vacuum manifolds
    • Allows easy, contamination-free stacking directly with collection plates
    • Protects recessed underdrain
    • Provides ample surface for bar code labels
    • Improves vacuum seal
  • Removable underdrain allows membrane access for ELISPOT assays, whole organism visualization, and radioisotope detection
  • Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
  • Opaque plastic prevents light transfer between wells during direct scintillation counting
  • Low binding hydrophilic Durapore™ membrane allows low nonspecific binding of antibodies, reagents, and samples
  • Meet ANSI™/SBS 2004 plate specifications
Description & Specifications

Specifications

Low
Clear
Receptor binding assays on whole cells and cell fragments, protein kinase, phosphatase precipitation assays, bead, resin based assays
Hydrophilic PVDF
0.22μm
Yes
0.05 to 0.25mL
ANSI, SBS
0.28cm2
Styrene
96
Pk. of 10
MultiScreenHTS-GV Plate