Please login to your online account to display your discounted pricing
EMD Millipore MultiScreenHTS Durapore™ 96-Well Filter Plates
Designed for full automation compatibility. EMD Millipore MultiScreenHTS Durapore™ 96-Well Filter Plates are innovative plates ideal for bead based assays. Opaque Barex™ and clear, sterile or non-sterile styrene and racks are available in a range of pore sizes.
Manufacturer: emd millipore  MSHVS4510
 View more versions of this product
Catalog No. MSHVS4510
  • / Pack of 10 Each
Description & Specifications
Specifications
Volume (Metric) Well 0.05 to 0.25mL
Filtration Area 0.28cm2
For Use With (Application) Receptor binding assays on whole cells and cell fragments, protein kinase, phosphatase precipitation assays, bead, resin based assays
Binding Property Low
Material Styrene
Membrane Hydrophilic PVDF
No. of Wells 96
Pore Size 0.45μm
Quantity Pk. of 10
Sterile Yes
Type MultiScreenHTS-HV Plate
Certifications/Compliance ANSI, SBS
Color Clear
Binding Property Low
For use in receptor-ligand binding, general sample preparation, ELISPOT, enzyme activity, bead-based assays, whole organism visualization, and radioisotope detection.
  • Standardized dimensions and aligned, rigid sidewalls
    • Allows full compatibility with automated/robotic gripper arms, stackers, plate washers, bar code labels, and plate readers
  • Unique two-piece plate design eliminates surface contact with individual well “drip directors” to prevent crosstalk between wells
  • Extended skirt and vacuum manifolds
    • Allows easy, contamination-free stacking directly with collection plates
    • Protects recessed underdrain
    • Provides ample surface for bar code labels
    • Improves vacuum seal
  • Removable underdrain allows membrane access for ELISPOT assays, whole organism visualization, and radioisotope detection
  • Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
  • Opaque plastic prevents light transfer between wells during direct scintillation counting
  • Low binding hydrophilic Durapore™ membrane allows low nonspecific binding of antibodies, reagents, and samples
  • Meet ANSI™/SBS 2004 plate specifications