Please login to your online account to display your discounted pricing

EMD Millipore MultiScreenHTS Durapore™ 96-Well Filter Plates


Designed for full automation compatibility. EMD Millipore MultiScreenHTS Durapore™ 96-Well Filter Plates are innovative plates ideal for bead based assays. Opaque Barex™ and clear, sterile or non-sterile styrene and racks are available in a range of pore sizes.

Manufacturer: emd millipore  MSDVN6510

 View more versions of this product

Catalog No. MSDVN6510

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Filtration Area 0.28cm2
For Use With (Application) Receptor binding assays on whole cells and cell fragments, protein kinase, phosphatase precipitation assays, bead, resin based assays
Binding Property Low
Material Styrene
Membrane Hydrophilic PVDF
No. of Wells 96
Pore Size 0.65μm
Quantity Pk. of 10
Sterile No
Type MultiScreenHTS-DV Plate
Certifications/Compliance ANSI, SBS
Volume (Metric) Well 0.05 to 0.25mL
Color Clear
Volume (Metric) Well 0.05 to 0.25mL
Binding Property Low

For use in receptor-ligand binding, general sample preparation, ELISPOT, enzyme activity, bead-based assays, whole organism visualization, and radioisotope detection.

  • Standardized dimensions and aligned, rigid sidewalls
    • Allows full compatibility with automated/robotic gripper arms, stackers, plate washers, bar code labels, and plate readers
  • Unique two-piece plate design eliminates surface contact with individual well “drip directors” to prevent crosstalk between wells
  • Extended skirt and vacuum manifolds
    • Allows easy, contamination-free stacking directly with collection plates
    • Protects recessed underdrain
    • Provides ample surface for bar code labels
    • Improves vacuum seal
  • Removable underdrain allows membrane access for ELISPOT assays, whole organism visualization, and radioisotope detection
  • Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
  • Opaque plastic prevents light transfer between wells during direct scintillation counting
  • Low binding hydrophilic Durapore™ membrane allows low nonspecific binding of antibodies, reagents, and samples
  • Meet ANSI™/SBS 2004 plate specifications