Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™ Solvinert Filter Plates


Optimized for drug discovery applications. EMD Millipore MultiScreen™ Solvinert Filter Plates are solvent compatible and feature a unique one-piece design. 96-well plates are available in deep well (2mL) format (with or without pre-filter) and standard (500μL) format, with hydrophobic or hydrophilic membranes.

Manufacturer: emd millipore  MDRPNP410

 View more versions of this product

Catalog No. MDRPNP410

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Type MultiScreen Deep-Well Solvinert Plate with Prefilter
No. per Pack 10/Pk.
Filtration Area 0.28cm2
For Use With (Application) Solid phase synthesis and cleanup, in plate protein precipitation, sample recovery following in-plate compound cleavage, peptide synthesis
Height (Metric) 4.07cm
Length (Metric) 1.28cm
Material Polyolefin Copolymer
Membrane Hydrophobic PTFE
No. of Wells 96
Pore Size 0.45μm
Sterile No
Volume (Metric) Well 1.9mL
Width (Metric) 8.55cm
Color Clear
Height (Metric) 4.07cm
Height (English) 1.6 in.
Length (English) 0.5 in.
Dimensions (L x W x H) 0.5 x 3.36 x 1.6 in. (1.28 x 8.55 x 4.07cm)
Volume (Metric) Well 1.9mL
Width (English) 3.36 in.

  • Clear polyolefin base plate offers broad solvent compatibility
  • One-piece design eliminates leaking and allows for extended incubations
  • Low binding and low extractables
  • Accommodate aqueous and nonaqueous samples with chemically resistant hydrophilic or hydrophobic membranes
  • Polytetrafluoroethylene membranes feature 45μL pores
  • Automation compatible
  • Available in 96-well deep well (2mL) format (with or without pre-filter) and standard (500μL) format