Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™ Sealing Tape


Choose from opaque or clear

Manufacturer: emd millipore  MATAH0P00

 View more versions of this product

Catalog No. MATAH0P00

  • / Each
 

Description & Specifications

Specifications

No. per Pack 100.0
Product Type Sealing Tape
Applications Used to seal 100μL plate bottoms read in TopCount™ instruments
Sterile No
Color Opaque