Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™ PCRμ96, PCR96, and PCR384 Filter Plates


Fully automatable PCR product purification

Manufacturer: emd millipore  S384PCR10

 View more versions of this product

Catalog No. S384PCR10

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Certifications/Compliance SBS
Format 384-well
Volume (Metric) Well 1 to 100μL
Material Styrene
Color Clear
Type Montage PCR384
No. of Wells 384
Sterile No
For Use With (Application) Purification of PCR products