Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™ HTS 96-Well Filter Plates with MCE Membrane


Commonly used for Elispot assays, bacterial screening, ELISA or viral infection studies

Manufacturer: emd millipore  MSHAS4B10

 View more versions of this product

Catalog No. MSHAS4B10

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Filtration Area 0.28cm2
For Use With (Application) Solid phase RIA, DNA hybridization, ELISA, protein binding
Material Barex™/TiO2
Membrane Mixed Cellulose Esters
No. of Wells 96
Pore Size 0.45μm
Quantity 10/Pk.
Sterile Yes
Type MultiScreenHTS-HA Plate
Color Opaque