Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™ HTS 96-Well Filter Plates with MCE Membrane


Commonly used for Elispot assays, bacterial screening, ELISA or viral infection studies

Manufacturer: emd millipore  MSHAN4550

 View more versions of this product

Catalog No. MSHAN4550

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Sterile No
Membrane Mixed Cellulose Esters
No. of Wells 96
Filtration Area 0.28cm2
For Use With (Application) Solid phase RIA, DNA hybridization, ELISA, protein binding
Type MultiScreenHTS-HA Filter Plate
Color Clear
Quantity 50/Pk.
Pore Size 0.45μm
Material Styrene

  • Developed for high-throughput use with automated work stations
  • Individually sealed wells prevent incubation crosstalk
  • Removable underdrain allows access to the filters
  • Rigid sidewalls improve handling
  • Ample surfaces provided for bar code labels
  • Pore size: .45μm

Solid-phase RIA, DNA hybridization, ELISA and protein binding