MilliporeSigma™ MultiScreen™ HTS 96-Well Filter Plates with MCE Membrane
Commonly used for Elispot assays, bacterial screening, ELISA or viral infection studies
$298.00 - $1,572.58
Specifications
Format Standard
Membrane Mixed Cellulose Esters
Array 8 x 12
Pore Size 0.45 μm
No. of Wells 96
View More Specs
Products 5
Catalog Number Mfr. No. Color Material Sterility Quantity Price Quantity  
Catalog Number Mfr. No. Color Material Sterility Quantity Price Quantity  
MSHAN4510
View Documents Promotion Details
MilliporeSigma™
MSHAN4510
Clear Styrene Non-sterile 10/Pk.
Pack of 10 Each for $298.00
MSHAN4550
View Documents Promotion Details
MilliporeSigma™
MSHAN4550
Clear Styrene Non-sterile 50/Pk.
Pack of 50 Each for $1,412.16
MSHAN4B50
View Documents Promotion Details
MilliporeSigma™
MSHAN4B50
Opaque Barex Non-sterile 50/Pk.
Pack of 50 Each for $1,572.58
MSHAS4510
View Documents Promotion Details
MilliporeSigma™
MSHAS4510
Clear Styrene Sterile 10/Pk.
Pack of 10 Each for $388.24
MSHAS4B10
View Documents Promotion Details
MilliporeSigma™
MSHAS4B10
Opaque Barex Sterile 10/Pk.
Pack of 10 Each for $403.00
Description
Description
  • Developed for high-throughput use with automated work stations
  • Individually sealed wells prevent incubation crosstalk
  • Removable underdrain allows access to the filters
  • Rigid sidewalls improve handling
  • Ample surfaces provided for bar code labels
  • Pore size: .45μm
Solid-phase RIA, DNA hybridization, ELISA and protein binding
Specifications
Specifications
Standard
8 x 12
96
Mixed Cellulose Esters
0.45 μm
Solid phase RIA, DNA hybridization, ELISA, protein binding
SDS