Please login to your online account to display your discounted pricing

EMD Millipore MultiScreenHTS 384-Well Filter Plates


Use in automated, high throughput applications, including biological assays and sample preparation. EMD Millipore MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.

Manufacturer: emd millipore  MZFCN0W10

 View more versions of this product

Catalog No. MZFCN0W10

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Membrane Glass Fiber, Polyester Mesh
Type MultiScreenHTS-FC Plate
Dimensions (L x W x H) 1.28 x 8.55 x 1.44cm
Material Barex™/TiO2
Volume (Metric) Well 0.02 to 0.1mL
For Use With (Application) Receptor, ligand binding assays, sample prep, TCA precipitation
Width (Metric) 8.55cm
No. of Wells 384
Color Opaque
Filtration Area 0.09cm2
Length (Metric) 1.28cm
Pore Size 1.2μm
Quantity 10
Height (Metric) 1.44cm
Sterile No
Height (Metric) 1.44cm
Volume (Metric) Well 0.02 to 0.1mL
Height (English) 0.56 in.
Length (English) 0.5 in.
Width (English) 3.36 in.

For use in receptor-ligand binding, general sample preparation, enzyme assays with precipitation and DNA purification.

  • Rigid sidewalls allows full compatibility with automated/robotic systems
  • Unique plate design and individually sealed filter discs prevent cross-talk between wells
  • Opaque plastic lids prevents light transfer between wells during direct scintillation counting
  • Barcoding available
  • Meet ANSI™/SBS standards