Please login to your online account to display your discounted pricing

EMD Millipore MultiScreenHTS 384-Well Filter Plates


Use in automated, high throughput applications, including biological assays and sample preparation. EMD Millipore MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.

Manufacturer: emd millipore  MZPHN0W10

 View more versions of this product

Catalog No. MZPHN0W10

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Dimensions (L x W x H) 1.28 x 8.55 x 1.44cm
Material Styrene Acrylonitrile (SAN)/TiO2
Type MultiScreenHTS-PH Plate
Filtration Area 0.09cm2
Height (Metric) 1.44cm
Volume (Metric) Well 0.02 to 0.1mL
For Use With (Application) Protein kinase assays with peptide substrates, enzyme assays, binds positive charge
Length (Metric) 1.28cm
Membrane Negatively Charged Phosphocellulose, Polyester Mesh
No. of Wells 384
Pore Size 0.65μm
Quantity 10
Sterile No
Width (Metric) 8.55cm
Color Opaque
Height (Metric) 1.44cm
Height (English) 0.56 in.
Length (English) 0.5 in.
Volume (Metric) Well 0.02 to 0.1mL
Width (English) 3.36 in.

For use in receptor-ligand binding, general sample preparation, enzyme assays with precipitation and DNA purification.

  • Rigid sidewalls allows full compatibility with automated/robotic systems
  • Unique plate design and individually sealed filter discs prevent cross-talk between wells
  • Opaque plastic lids prevents light transfer between wells during direct scintillation counting
  • Barcoding available
  • Meet ANSI™/SBS standards