Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™ High Volume Filter Plates


Multiwell solutions for discovery research and sample prep

Manufacturer: emd millipore  MVFCN1225

 View more versions of this product

Catalog No. MVFCN1225

  • / Pack of 25 Each
 

Description & Specifications

Specifications

No. of Wells 96
Width (English) 3.37 in.
Color Opaque
Pore Size 1.2μm
Dimensions (L x W x H) 0.5 x 3.37 x 1.85 in. (1.27 x 8.56 x 4.7cm)
Width (Metric) 8.56cm
Length (English) 0.5 in.
Material Polypropylene
Type MultiScreen High Volume FC Plate
Length (Metric) 1.27cm
For Use With (Application) Receptor, ligand binding assays, sample prep, TCA precipitation
Height (Metric) 4.7cm
Sterile No
Height (English) 1.85 in.
Volume (Metric) Well 2mL