missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore™ MultiScreen™ Harvest Plates

This Opaque 96-well Cell Harvesting Plates has a Glass Fiber membrane used for Cell harvesting.

Manufacturer:  EMD Millipore™ MAHFC1H60

 View more versions of this product

Catalog No. MAHFC1H60



Description

Description

Opaque plates feature choice of Millipore glass fiber B (FB) or glass fiber C (FC) filter media and promote high and uniform flowrates in standard 96-well plate cell harvesters and readers.
•    Made of Barex/TiO2-filled SAN (styrene acrylonitrile) that is resistant to all aqueous solutions including moderate-to-strong acids; fully compatible with standard biosafe scintillants
•    B1 and C1 plates have 100μL well capacity
•    Optimal % open area (for light transmission in coincidence counting) of 26% (in 100μL plates)
•    Filters have an effective filtration area of 0.33cm2 and a pore size of 1.0μm for FB or 1.2μm for FC types
•    Nonsterile; without lid

Order Info

Pack of 60 (in 6 sleeves of 10 plates each)

Compliance

Each lot is quality checked for uniform flowrate and full particulate retention and meets all specifications for SBS Level 3

Specifications

Specifications

SBS
Cell harvesting
96
0.1mL
Non-sterile
Opaque
Barex&trade
1.2μm
Multiscreen Harvest-C1; Glass fiber FC
Documents