Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™ Harvest Plates


Manufacturer: emd millipore  MAHFB1H60

 View more versions of this product

Catalog No. MAHFB1H60

  • / Pack of 60 Each
 

Description & Specifications

Specifications

Volume (Metric) Well 0.1mL
Pore Size 1.0μm
Type Multiscreen Harvest-B1; Glass fiber FB
Color Opaque
No. of Wells 96
For Use With (Application) Cell harvesting
Sterile No
Certifications/Compliance SBS
Material Barex™/TiO2