Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™ Cell Culture Trays


For feeding cells during membrane-based cell culture

Manufacturer: emd millipore  MAMCS0110

 View more versions of this product

Catalog No. MAMCS0110

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Lid No
No. of Wells 1
Format 1-well
Sterile Yes
Closure No
Color Clear
Material Polystyrene
Type MultiScreen Single Well Culture Tray
For Use With (Application) Cell culture applications, use with sterile MultiScreen filtration plate