EMD Millipore MultiScreen™ Cell Culture Trays
For feeding cells during membrane-based cell culture
$125.00 - $203.00
Specifications
Color Clear
Sterile Yes
Lid No
Material Polystyrene
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) No. of Wells Format Type Price Quantity  
Catalog Number Mfr. No. For Use With (Application) No. of Wells Format Type Price Quantity  
MAMCS0110 EMD Millipore
MAMCS0110
Cell culture applications, use with sterile MultiScreen filtration plate 1 1-well MultiScreen Single Well Culture Tray Pack of 10 Each for $125.00
MAMCS9610 EMD Millipore
MAMCS9610
Cell transport assays, use with sterile MultiScreen filtration plate 96 96-well MultiScreen 96-Well Culture Tray Pack of 10 Each for $203.00
Specifications
Description
For basolateral feeding and/or co-culture in a single well or 96-well format
  • Use with sterilized MultiScreen filter plates
  • Polystyrene with cut-off corners
  • Sterilized and individually wrapped
  • Untreated surfaces
Single-Well Tray
  • Holds medium and commonly feeds all 96 wells
  • 96-Well Tray
    • Independent wells fit the 96 wells of the membrane plate
    • Patented “tear drop” shaped wells prevent air entrapment under membrane
    Description & Specifications
    Specifications
    Clear
    Yes
    No
    Polystyrene