EMD Millipore™ MultiScreen™ Carrier Racks
For radionucleotide counting by transfer of individual filters to tubes
Manufacturer:  EMD Millipore™ MACR08124
 View more versions of this product
Catalog No. MACR08124
Description
Description
  • Carrier racks hold sample collection containers in place; available for 4mL vials, 7mL vials, or 12 x 75mm tubes (12 racks per carrier rack assembly)
Specifications
Specifications
Radionucleotide counting by transfer of individual filters to tubes
Carrier Rack Assemblies for 4mL Vials
Documents