missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ MultiScreen™ 96-Well Assay Systems

Perform cell growth, feeding, and analysis in one plate

Manufacturer:  MilliporeSigma™ PSRP004R5

 View more versions of this product

Catalog No. PSRP004R5



Description

Description

  • Patented 96-well device supports epithelial cell growth and differentiation of the Caco-2 and other cell lines
  • After the formation of a differentiated Caco-2 monolayer, the assembly can be used to measure the rate of drug transport across the Caco-2 cell barrier
  • All procedures are designed to be carried out in a single device
  • Apical and basolateral access ports allow contamination-free access to cells, easy feeding and media changes, and effective sample analysis
  • Increased sample throughput when compared to 24-well systems
  • Use manually or with automated systems
  • Versatile format with 96- or single-well feeder trays

Specifications

Specifications

Clear
0.12 cm2
96-well cell culture plate, single well feeder tray, 96-well receiver tray, lid
Polystyrene
1 μm
2.62 cm
Bulk
Sterile
With Lid
8 x 12
0.5 x 3.33 x 1.03 in. (1.28 x 8.48 x 2.62 cm)
Cell attachment, growth and differentiation for many cell applications
1.28 cm
Polyethylene Terephthalate
5/Pk.
8.48 cm
Millicell 96 Cell Culture Plate
Standard
96
Documents