Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™ 96-Well Assay Plates for Sample Preparation


For the isolation, purification, and preparation of samples prior to testing in drug discovery processes and protein and DNA applications

Manufacturer: emd millipore  MAIPN0B50

 View more versions of this product

Catalog No. MAIPN0B50

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Filtration Area 0.28cm2
For Use With (Application) Protein interaction assays, avidin biotin linkages, DNA binding proteins
Material Barex™/TiO2
Membrane Hydrophobic PVDF
No. of Wells 96
Pore Size 0.45μm
Sterile No
Type MultiScreen-IP Filter Plate
Volume (Metric) Well 0.05 to 0.25mL
Width (English) 3.36 in.
Width (Metric) 8.55cm
Color Opaque
Volume (Metric) Well 0.05 to 0.25mL