Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™ 96-Well Assay Plates for ELISPOT Assays


Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining

Manufacturer: emd millipore  MSIPN4W50

 View more versions of this product

Catalog No. MSIPN4W50

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Filtration Area 0.28cm2
For Use With (Application) ELISPOT assays
Material Acrylic
Membrane Hydrophobic PVDF
No. of Wells 96
Pore Size 0.45μm
Sterile No
Type MultiScreenHTS-IP Non-Sterile Filter Plate
Color White