EMD Millipore™ MultiScreen™ 96-Well Assay Plates for Cell-Based Assays
Offer formats that allow a wide range of functional cell-based and whole-organism assays to be performed
Manufacturer:  EMD Millipore™ MAMIC8S10
 View more versions of this product
Catalog No. MAMIC8S10
Description
Description
Ideal for cell viability and proliferation, migration, invasion, and chemotaxis assays. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.
Specifications
Specifications
Clear
Migration, invasion and chemotaxis
Polycarbonate
8μm
Sterile
0.28cm2
Styrene
96
MultiScreen-MIC Plate
Documents