Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™ 96-Well Assay Plates


Provide tools to perform high-throughput studies of ADME processes—the investigation of the adsorption, distribution, metabolism, and excretion of potential drugs

Manufacturer: emd millipore  MSSACCEPT0R

 View more versions of this product

Catalog No. MSSACCEPT0R

  • / Pack
 

Description & Specifications

Specifications

For Use With (Application) Permeability and PAMPA assays
Material PTFE
Sterile No
Type MultiScreen Acceptor Plate
Color White