Please login to your online account to display your discounted pricing

EMD Millipore MultiScreen™ 96-Well Assay Plates


Provide tools to perform high-throughput studies of ADME processes—the investigation of the adsorption, distribution, metabolism, and excretion of potential drugs

Manufacturer: emd millipore  MPC4NTR10

 View more versions of this product

Catalog No. MPC4NTR10

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Pore Size 0.45μm
Sterile No
No. of Wells 96
Color Clear
Material Styrene
For Use With (Application) Permeability studies
Type MultiScreen Permeability