Please login to your online account to display your discounted pricing

EMD Millipore MQuant™ Nitrite Test Strips


To determine nitrite in analysis of water and cooling lubricants

Manufacturer: emd millipore  1100570001

 View more versions of this product

Catalog No. M1100570001

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) To determine nitrite in analysis of water and cooling lubricants
Graduations 0.5-1-2-5-10mg/L
For Use With (Equipment) Colorimeter
No. of Tests 75.0
Storage Requirements 2° to 8°C
Method Griess' reaction
Product Type Test Strip
Range 0.5 to 10mg/L NO2-