Please login to your online account to display your discounted pricing

EMD Millipore MQuant™ Nitrate Test Strips


To determine nitrate in aqueous media and food

Manufacturer: emd millipore  1100200001

 View more versions of this product

Catalog No. M1100200001

  • / Each
 

Description & Specifications

Specifications

No. of Tests 100.0
Method Modified Griess' reaction
For Use With (Application) For food and water analysis
Product Type Test Strip
For Use With (Equipment) Colorimeter
Storage Requirements 2° to 8°C
Range 10 to 500mg/L
Graduations 10-25-50-100-250-500mg/L