missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ MQuant™ Nitrate Test Strips
To determine nitrate in aqueous media and food
Supplier: MilliporeSigma™ 1100920021
Includes: Tube containing 100 strips
Description
- Small and easy to handle
- Flexible and universal application
- Modified Griess' reaction method ideal for screening
- Immediate result produced through on-site analysis
- PET film and low reagent content
- Easily disposable
- Color-scale graduation: 10 - 25 - 50 - 100 - 250 - 500mg/L
Reclose the tube containing the test strips immediately after use.
For food and water analysis
Specifications
For food and water analysis | |
10-25-50-100-250-500mg/L | |
10 to 500mg/L NO3- | |
Modified Griess' reaction |
Colorimeter | |
1000 | |
2°C to 8°C | |
Test Strip |