Please login to your online account to display your discounted pricing

EMD Millipore MQuant™ Chlorine Test Strips


To determine the presence of residues of chlorine

Manufacturer: emd millipore  1179240001

 View more versions of this product

Catalog No. M1179240001

  • / Each
 

Description & Specifications

Specifications

No. of Tests 100.0
For Use With (Application) For water analysis, rapid determination of higher concentrations of chlorinated disinfectants, which are chiefly used in the food industry
Graduations 25-50-100-200-500mg/L
For Use With (Equipment) Colorimeter
Storage Requirements 2° to 8°C
Method Redox Reaction
Product Type Test Strip
Range 25 to 500mg/L