missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore™ MQuant™ Arsenic Test Strips

More secure and easy to use

$553.51 - $553.51

Specifications

Product Type MQuant™ Arsenic Test Strip
For Use With (Application) For measuring arsenic in water, soil extracts, pharmaceutical products, prepared biological specimens and liquid foods
Quantity 100 Tests
Storage Requirements 15° to 25°C
Test Method Modified Gulzeit test
View More Specs

Includes: Reagent

Products 1
Catalog Number Mfr. No. Graduations Includes Range Price Quantity  
Catalog Number Mfr. No. Graduations Includes Range Price Quantity  
M1179170001 View Documents EMD Millipore™
M1179170001
0.02-0.05-0.1-0.2-0.5mg/L 2 bottles of reagent As-1, 1 bottle of reagent As-2, 1 graduated 5-ml plastic syringe, 1 red dosing spoon, 2 reaction tubes with screw caps 0.02 to 3mg/L Each for $553.51
Description

Description

Comfortable and secure transport for on-site analyses.

  • Designed for upright transport of reagents
  • Solid acid is used as a reagent to avoid transporting liquid acid and zinc
  • Two reagent bottles to conduct two tests simultaneously
  • Modified Gutzeit test
  • PET film and low reagent content for easier disposal
  • Small and easy to handle for on-site use
  • Fast results
  • Ideal for screening

  • Available in practical case (M1179270001) or stable plastic box (M1179170001)
  • M1179170001 Graduations: 0.02-0.05-0.1-0.2-0.5mg/L; 0.1-0.5-1.0-1.7-3mg/L
  • M1179270001 Graduations: 0.005-0.01-0.025-0.05-0.1-0.25-0.5mg/L

Specifications

Specifications

MQuant™ Arsenic Test Strip
100 Tests
Modified Gulzeit test
For measuring arsenic in water, soil extracts, pharmaceutical products, prepared biological specimens and liquid foods
15° to 25°C