Please login to your online account to display your discounted pricing
EMD Millipore Montage™ SEQ384 Sequencing Reaction Cleanup Kit
Use 384-well filter plates with proprietary size-exclusion membranes to retain sequencing products while excess dye terminator is removed quickly and efficiently. Fully automatable kits rinse away unincorporated dye terminator without the need for binding and eluting protocols.
Manufacturer: emd millipore  S384SEQ50
 View more versions of this product
MultiScreen™ 384 Vacuum Manifold with underdrain support grid (Mfr. No. MAVM 096 0R)
Catalog No. S384SEQ50
  • / Pack of 50 Each
Description & Specifications
Specifications
For Use With (Application) DNA sequencing
Material Polystyrene
Includes Clear Polystyrene 384-well plates
Quantity 50 Pack
Product Name Kit
Quantity 50 Pack
  • Recovered sequencing products are highly purified and result in long, accurate sequencing reads
  • Protocols are optimized for use with BigDye™ Terminator version 3.0, 3.1, 1.1 and ET Terminator sequencing chemistries
  • Sequencing products may be resuspended in any aqueous electrokinetic injection buffer
  • Plates are suitable for use with 1/8 sequencing reactions, reducing reagent costs by miniaturization