Please login to your online account to display your discounted pricing

EMD Millipore Montage™ Plasmid Miniprep96 Kits


Generate high-quality plasmid DNA in 50% less time than traditional methods

Manufacturer: emd millipore  LSKP09624

 View more versions of this product

MultiScreen™ Vacuum Manifold with underdrain support grid (MAVM 096 0R)

Catalog No. LSKP09624

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Plasmid Miniprep, BAC Prep
Format Microplate
Includes All necessary reagents and disposables
Product Type Kit
Quantity 24 x 96 Preps
Product Name Kit
Quantity 24 x 96 Preps