missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Montage™ Plasmid Miniprep96 Kits

Generate high-quality plasmid DNA in 50% less time than traditional methods

$939.00 - $4,860.00

Specifications

Format Microplate
For Use With (Application) Plasmid Miniprep, BAC Prep
Includes All necessary reagents and disposables
View More Specs

Includes: All necessary reagents and disposables

Requires: MultiScreen™ Vacuum Manifold with underdrain support grid (MAVM 096 0R)


Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LSKP09624 View Documents MilliporeSigma™
LSKP09624
24 x 96 Preps Each for $4,860.00
 
LSKP09604 View Documents MilliporeSigma™
LSKP09604
4 x 96 Preps Each for $939.00
 
Description

Description

  • Typical yield from 1mL of bacterial culture is 4 to 6μg
  • 96-well kit uses unique separation technology Simple, 25-minute protocol yields clean and reproducible DNA for sensitive downstream applications Collection of purified sample from the top of the plate and lack of centrifugation steps allow compatibility with automated systems

Specifications

Specifications

Microplate
Plasmid Miniprep, BAC Prep
All necessary reagents and disposables