Please login to your online account to display your discounted pricing

EMD Millipore Montage™ Antibody Purification Kits with Prosep™-A and Prosep-G Media


Provide a centrifugal method for fast and easy purification of antibodies from serum, ascites, or cell culture supernatants

Manufacturer: emd millipore  LSK2ABG60

 View more versions of this product

Catalog No. LSK2ABG60

  • / Each
 

Description & Specifications

Specifications

Quantity 6 Pack
Support Type Porous Glass
Color Code Yellow
For Use With (Application) Antibody purification
Sterile No
Storage Requirements 2° to 8°C
Volume (Metric) Bed 1600μL