Please login to your online account to display your discounted pricing

EMD Millipore Montage™ Antibody Purification Kits with Prosep™-A and Prosep-G Media


Provide a centrifugal method for fast and easy purification of antibodies from serum, ascites, or cell culture supernatants

Manufacturer: emd millipore  LSK2ABG20

 View more versions of this product

Catalog No. LSK2ABG20

  • / Each
 

Description & Specifications

Specifications

Volume (Metric) 20mL
Color Code Yellow
For Use With (Application) Antibody purification
Volume (English) 0.67 oz.
Volume (Metric) Bed 1600μL
Target Antibody
Sterile No
Storage Requirements 2° to 8°C
Support Type Porous Glass
Quantity 2 Pack