Please login to your online account to display your discounted pricing

EMD Millipore Montage™ Antibody Purification Kits with Prosep™-A and Prosep-G Media

Provide a centrifugal method for fast and easy purification of antibodies from serum, ascites, or cell culture supernatants

Manufacturer: emd millipore  LSK2ABA20

 View all options for this product

Catalog No. LSK2ABA20


Description & Specifications


Quantity 2 Pack
Support Type Porous Glass
Target Antibody
Color Code Red
For Use With (Application) Antibody purification
Volume (Metric) 20mL
Volume (Metric) Bed 1600μL
Volume (English) 0.67 oz.
Sterile No
Storage Requirements 2° to 8°C

The Prosep family of media has a long history of success for large-scale bioprocessing applications including Prosep-A for industrial monoclonal antibody purification. Montage Kits deliver that same level of performance and reliability for research-scale antibody purification.

  • Purify up to 10mL of sample in less than 60 minutes Yield up to 20mg of purified antibody
  • Utilize Prosep high-capacity media, which offers rapid transport and a very high dynamic capacity for IgG