missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore Montage™ Antibody Purification Kits with Prosep™-A and Prosep-G Media

Provide a centrifugal method for fast and easy purification of antibodies from serum, ascites, or cell culture supernatants

$1,139.00 - $1,221.00

Specifications

For Use With (Application) Antibody purification
Sterile No
Storage Requirements 2° to 8°C
Support Type Porous Glass
Volume (Metric) Bed 1600μL
View More Specs

Includes:
  • LSK2ABA20 and LSK2ABG20: 2 Media plugs, 2 Spin columns, 4 Centrifuge tubes, 4 Steriflip™ Devices, 4 Amicon™ Ultra Centrifugal Devices, Optimized buffers


Products ${productFamilyLength}
Catalog Number Mfr. No. Color Code Quantity Price Quantity  
Catalog Number Mfr. No. Color Code Quantity Price Quantity  
LSK2ABA60 EMD Millipore
LSK2ABA60
Red 6 Pack Each for $1,139.00
LSK2ABG60 EMD Millipore
LSK2ABG60
Yellow 6 Pack Each for $1,221.00
Specifications

Description

The Prosep family of media has a long history of success for large-scale bioprocessing applications including Prosep-A for industrial monoclonal antibody purification. Montage Kits deliver that same level of performance and reliability for research-scale antibody purification.

  • Purify up to 10mL of sample in less than 60 minutes Yield up to 20mg of purified antibody
  • Utilize Prosep high-capacity media, which offers rapid transport and a very high dynamic capacity for IgG

Order Info

Montage Spin Columns with Prosep media plugs are also available separately in packs of six. Each column can perform 10 purifications.

Description & Specifications

Specifications

Antibody purification
2° to 8°C
1600μL
No
Porous Glass