missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Mitex™ PTFE Membrane Filters

Unlaminated for extreme chemical or temperature conditions

Manufacturer:  MilliporeSigma™ LCWP14250

 View more versions of this product

Catalog No. LCWP14250



Description

Description

  • Membranes have no backing
  • Mean thickness is 125μm
  • Autoclavable at 121°C (250°F)

Applications:

Clarification of acids, bases and cryogenic fluids; clarification of propellants; analysis of hydraulic fluids; RNA isolation.

Specifications

Specifications

10μm
142mm
50/pk.
0.5psi
125μm
Polytetrafluoroethylene (PTFE)
White
Yes
Clarification of acids, bases, and cryogenic fluids; Clarification of propellants; Analysis of hydraulic fluids; RNA isolation.
Plain
Documents
Promotions

Promotions